siRNA Details
Gene Symbol: ZNF789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213603.2 | NP_998768.2 | 5 | 1570 | ||
XM_011516067.2 | XP_011514369.1 | 6 | 1726 | |||
XM_011516070.2 | XP_011514372.1 | 4 | 1564 | |||
XM_011516073.2 | XP_011514375.1 | 3 | 1329 | |||
XM_017012018.1 | XP_016867507.1 | 5 | 1522 | |||
XM_017012019.1 | XP_016867508.1 | 5 | 5638 | |||
XM_017012020.1 | XP_016867509.1 | 6 | 5791 | |||
XM_017012021.1 | XP_016867510.1 | 7 | 4095 | |||
XM_017012022.1 | XP_016867511.1 | 3 | 1408 | |||
XM_017012023.1 | XP_016867512.1 | 4 | 1528 | |||
XM_017012024.1 | XP_016867513.1 | 5 | 3506 | |||
XM_017012025.1 | XP_016867514.1 | 6 | 5799 | |||
XM_017012026.1 | XP_016867515.1 | 6 | 1801 | |||
XM_017012027.1 | XP_016867516.1 | 7 | 1884 | |||
GenBank mRNA | AK131429.1 | - | 1 | 1134 | ||
BC089424.1 | - | 5 | 1532 | 3054018 | ||
BX648360.1 | - | 3 | 2158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
