siRNA Details
Gene Symbol: OR1J1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004451.1 | NP_001004451.1 | 1 | 583 | ||
GenBank mRNA | BC136933.1 | - | 1 | 613 | 9020925 | |
BC136938.1 | - | 1 | 613 | 9020930 |
