siRNA Details
Gene Symbol: OR2AE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005276.1 | NP_001005276.1 | 1 | 841 | ||
GenBank mRNA | BC137375.1 | - | 1 | 865 | 9021373 | |
BC137376.1 | - | 1 | 865 | 9021374 |
