siRNA Details
Gene Symbol: OR2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004685.1 | NP_001004685.1 | 1 | 109 | ||
GenBank mRNA | BC136812.1 | - | 1 | 176 | 9020802 | |
BC136813.1 | - | 1 | 176 | 9020803 |
