siRNA Details
Gene Symbol: OR6F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005286.1 | NP_001005286.1 | 1 | 893 | ||
GenBank mRNA | BC137142.1 | - | 1 | 941 | 9021139 | |
BC137143.1 | - | 1 | 941 | 9021140 |
