siRNA Details
Gene Symbol: OR6M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005325.1 | NP_001005325.1 | 1 | 496 | ||
GenBank mRNA | BC136929.1 | - | 1 | 534 | 9020921 | |
BC136931.1 | - | 1 | 534 | 9020923 |
