siRNA Details
Gene Symbol: OR6V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001667.1 | NP_001001667.1 | 1 | 760 | ||
GenBank mRNA | BC137044.1 | - | 1 | 781 | 9021040 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR6V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001667.1 | NP_001001667.1 | 1 | 760 | ||
GenBank mRNA | BC137044.1 | - | 1 | 781 | 9021040 |