siRNA Details
Gene Symbol: OR8D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002917.1 | NP_001002917.1 | 1 | 202 | ||
GenBank mRNA | AF238489.1 | - | 1 | 202 | ||
BC136950.1 | - | 1 | 273 | 9020942 | ||
BC136954.1 | - | 1 | 273 | 9020946 |
