siRNA Details
Gene Symbol: SPRR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005988.2 | NP_005979.1 | 2 | 370 |
Gene Symbol: SPRR2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024209.3 | NP_001019380.2 | 2 | 373 | ||
GenBank mRNA | BC058018.1 | - | 2 | 388 | ||
DA761491.1 | - | 2 | 372 | |||
M20030.1 | - | 2 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
