siRNA Details
Gene Symbol: ZNF805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023563.3 | NP_001018857.2 | 4 | 1200 | ||
NM_001145078.1 | NP_001138550.1 | 3 | 1073 | |||
GenBank mRNA | AF024708.1 | - | 1 | 87 | ||
AK297977.1 | - | 3 | 1092 | |||
BC140786.1 | - | 1 | 339 | 9021654 | ||
CR936617.1 | - | 1 | 378 |
Gene Symbol: ZNF543 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213598.3 | NP_998763.2 | 4 | 1338 | ||
GenBank mRNA | AF024696.1 | - | 1 | 96 | ||
AK131547.1 | - | 4 | 1326 | |||
AL834534.1 | - | 3 | 907 | |||
BC101012.2 | - | 4 | 981 | 40006636 | ||
BC101013.1 | - | 4 | 981 | 40006637 | ||
BC101014.2 | - | 4 | 981 | 40006638 | ||
BC101015.1 | - | 4 | 981 | 40006640 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
