siRNA Details
Gene Symbol: GRHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198182.2 | NP_937825.2 | 5 | 899 | ||
XM_006711882.2 | XP_006711945.1 | 5 | 899 | |||
XM_011510343.2 | XP_011508645.1 | 5 | 870 | |||
XM_017003900.1 | XP_016859389.1 | 5 | 972 | |||
XM_017003901.1 | XP_016859390.1 | 5 | 972 | |||
GenBank mRNA | AF198489.1 | - | 5 | 837 | ||
AK296982.1 | - | 5 | 977 | |||
AK312950.1 | - | 5 | 869 | |||
BC067519.1 | - | 5 | 780 | 6971723 | ||
BC067520.1 | - | 5 | 780 | 6971724 | ||
BC067521.1 | - | 5 | 780 | 6971725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
