siRNA Details
Gene Symbol: FAM92A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283034.1 | NP_001269963.1 | 3 | 441 | ||
NM_145269.4 | NP_660312.2 | 3 | 441 | |||
NR_104267.1 | 3 | 441 | ||||
NR_104268.1 | 3 | 441 | ||||
XM_005250787.2 | XP_005250844.1 | 3 | 311 | |||
GenBank mRNA | AK096298.1 | - | 3 | 335 | ||
BC014598.2 | - | 3 | 441 | |||
BC108707.1 | - | 3 | 403 | |||
CR627475.1 | - | 3 | 1480 | |||
DQ327715.1 | - | 3 | 355 | |||
DQ327716.1 | - | 3 | 355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
