siRNA Details
Gene Symbol: GNRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000406.2 | NP_000397.1 | 2 | 2295 | ||
GenBank mRNA | AY392011.1 | - | 2 | 544 | ||
BC113546.1 | - | 2 | 570 | 8322598 | ||
L03380.1 | - | 2 | 599 | |||
L07949.1 | - | 2 | 568 | |||
S60587.1 | - | 2 | 568 | |||
S77472.1 | - | 2 | 569 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
