siRNA Details
Gene Symbol: LOC400682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528477.2 | XP_011526779.2 | 4 | 1568 | ||
GenBank mRNA | BX647610.1 | - | 2 | 1369 |
Gene Symbol: ZNF100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173531.3 | NP_775802.2 | 5 | 2259 | ||
GenBank mRNA | AK126551.1 | - | 5 | 2210 | ||
BC031248.1 | - | 4 | 2224 | |||
BC035579.1 | - | 5 | 2166 | 5576923 | ||
BX640982.1 | - | 1 | 969 |
