siRNA Details
Gene Symbol: GANAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278192.1 | NP_001265121.1 | 3 | 366 | ||
NM_198335.3 | NP_938149.2 | 6 | 708 | |||
GenBank mRNA | AF144074.1 | - | 6 | 597 | ||
AK302752.1 | - | 3 | 272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
