siRNA Details
Gene Symbol: VWA5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130142.1 | NP_001123614.1 | 19 | 2865 | ||
NM_014622.4 | NP_055437.2 | 18 | 2750 | |||
XM_011542828.2 | XP_011541130.1 | 19 | 2976 | |||
GenBank mRNA | AF002672.1 | - | 17 | 2662 | ||
AK291698.1 | - | 18 | 2704 | |||
AY366504.1 | - | 9 | 1327 | |||
AY366505.1 | - | 18 | 2750 | |||
AY366507.1 | - | 10 | 1442 | |||
AY366508.1 | - | 19 | 2865 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
