siRNA Details
Gene Symbol: TSPY26P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002781.1 | 1 | 519 | |||
GenBank mRNA | BC101556.1 | - | 1 | 519 | ||
BC101558.1 | - | 1 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
