siRNA Details
Gene Symbol: LRRC70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181506.4 | NP_852607.3 | 2 | 741 | ||
GenBank mRNA | AK123022.1 | - | 2 | 741 | ||
AY280614.1 | - | 2 | 586 | |||
AY358733.1 | - | 2 | 605 | |||
BC067275.1 | - | 1 | 270 | |||
BC110838.1 | - | 2 | 593 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
