siRNA Details
Gene Symbol: KCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199349.2 | NP_955381.2 | 21 | 2437 | ||
GenBank mRNA | AK095464.1 | - | 14 | 1911 | ||
AK122706.1 | - | 21 | 2437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
