siRNA Details
Gene Symbol: CLNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052964.2 | NP_443196.2 | 3 | 182 | ||
XM_011513775.2 | XP_011512077.1 | 3 | 413 | |||
XM_017007684.1 | XP_016863173.1 | 3 | 413 | |||
XM_017007685.1 | XP_016863174.1 | 3 | 413 | |||
GenBank mRNA | AB110420.1 | - | 3 | 285 | ||
AK093001.1 | - | 3 | 184 | |||
BC029887.1 | - | 3 | 212 | |||
BC137553.1 | - | 3 | 111 | 9021553 | ||
BC137556.1 | - | 3 | 111 | 9021556 |
