siRNA Details
Gene Symbol: Rps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005528.1 | NP_001005528.1 | 3, 4 | 314 | ||
GenBank mRNA | BC103658.1 | - | 3, 4 | 319 | 7446085 | |
FQ210084.1 | - | 3, 4 | 332 | |||
FQ217008.1 | - | 3, 4 | 320 | |||
FQ217453.1 | - | 3, 4 | 304 | |||
FQ221194.1 | - | 3, 4 | 335 | |||
FQ221611.1 | - | 3, 4 | 334 | |||
FQ221873.1 | - | 3, 4 | 333 | |||
FQ228420.1 | - | 3, 4 | 335 |
