siRNA Details
Gene Symbol: Pla2g6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005560.1 | NP_001005560.1 | 2 | 161 | ||
NM_001270796.1 | NP_001257725.1 | 3 | 240 | |||
XM_006241997.2 | XP_006242059.1 | 2 | 983 | |||
XM_006241998.2 | XP_006242060.1 | 2 | 937 | |||
XM_006241999.2 | XP_006242061.1 | 2 | 983 | |||
XM_006242000.2 | XP_006242062.1 | 2 | 662 | |||
XM_006242001.2 | XP_006242063.1 | 3 | 294 | |||
XM_006242002.1 | XP_006242064.1 | 2 | 530 | |||
XM_006242003.1 | XP_006242065.1 | 3 | 602 | |||
XM_006242004.2 | XP_006242066.1 | 2 | 223 | |||
XM_006242005.2 | XP_006242067.1 | 2 | 983 | |||
XM_006242006.2 | XP_006242068.1 | 2 | 661 | |||
XM_008765756.1 | XP_008763978.1 | 3 | 570 | |||
XM_008765757.1 | XP_008763979.1 | 2 | 983 | |||
GenBank mRNA | BC081916.1 | - | 2 | 161 | 7112407 |
