siRNA Details
Gene Symbol: SORCS3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104028.1 | 1 | 645 | |||
GenBank mRNA | AK097854.1 | - | 1 | 645 | ||
BC042079.1 | - | 1 | 629 | |||
BU852305.1 | - | 1 | 648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
