siRNA Details
Gene Symbol: Kars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006967.1 | NP_001006968.1 | 10 | 1277 | ||
XM_006255630.1 | XP_006255692.1 | 9 | 1149 | |||
GenBank mRNA | BC083652.1 | - | 10 | 1277 | 7135143 |
