siRNA Details
Gene Symbol: Uxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006982.1 | NP_001006983.1 | 2 | 146 | ||
XM_006256611.2 | XP_006256673.1 | 3 | 203 | |||
GenBank mRNA | BC082752.1 | - | 2 | 146 | 7303683 | |
FQ210799.1 | - | 3 | 191 |
