siRNA Details
Gene Symbol: Tuba4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007004.1 | NP_001007005.1 | 4 | 710 | ||
XM_006245227.2 | XP_006245289.1 | 4 | 1109 | |||
GenBank mRNA | BC083726.1 | - | 4 | 710 | 7191229 | |
FQ212913.1 | - | 734 | ||||
FQ212965.1 | - | 4 | 734 | |||
FQ213109.1 | - | 4 | 734 | |||
FQ213658.1 | - | 4 | 734 | |||
FQ213871.1 | - | 4 | 734 | |||
FQ215031.1 | - | 4 | 734 | |||
FQ215251.1 | - | 4 | 734 | |||
FQ218669.1 | - | 4 | 734 | |||
FQ225881.1 | - | 4 | 734 | |||
FQ225974.1 | - | 4 | 734 | |||
FQ232744.1 | - | 4 | 734 | |||
FQ232854.1 | - | 4 | 731 | |||
FQ234507.1 | - | 4 | 734 | |||
FQ235001.1 | - | 4 | 734 |

More Information
Other Species Targeted

Gene Ontology Categories:
