siRNA Details
Gene Symbol: Arhgdia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007005.1 | NP_001007006.1 | 6 | 1153 | ||
XM_006247897.2 | XP_006247959.1 | 6 | 1287 | |||
XM_006247898.2 | XP_006247960.1 | 6 | 1236 | |||
GenBank mRNA | AF506037.2 | - | 1 | 328 | ||
BC083817.1 | - | 6 | 1153 | 7110945 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133796, AK172140, AK077343, AK188655, AK170553, and others.
Gene Ontology Categories:
