siRNA Details
Gene Symbol: Rnaseh2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007007.1 | NP_001007008.1 | 9, 10 | 735 | ||
XM_008770762.1 | XP_008768984.1 | 9, 10 | 2808 | |||
XM_008770763.1 | XP_008768985.1 | 9, 10 | 2808 | |||
GenBank mRNA | BC083791.1 | - | 9, 10 | 735 | 7106440 | |
FQ220108.1 | - | 9, 10 | 762 |
