siRNA Details
Gene Symbol: Zpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007011.1 | NP_001007012.1 | 3 | 303 | ||
XM_006247384.2 | XP_006247446.1 | 3 | 1433 | |||
XM_006247385.2 | XP_006247447.1 | 3 | 1360 | |||
XM_006247387.2 | XP_006247449.1 | 3 | 1526 | |||
GenBank mRNA | AY251605.1 | - | 3 | 295 | ||
AY251606.1 | - | 2 | 229 | |||
BC082007.1 | - | 3 | 303 | 7131668 |
