siRNA Details
Gene Symbol: Rbm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007014.2 | NP_001007015.2 | 4 | 1585 | ||
XM_008760164.1 | XP_008758386.1 | 3 | 4440 | |||
XM_008760165.1 | XP_008758387.1 | 3 | 4429 | |||
XM_008760166.1 | XP_008758388.1 | 4 | 1828 | |||
GenBank mRNA | BC086416.1 | - | 4 | 1585 | 7130401 |
