siRNA Details
Gene Symbol: Ctnna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007145.1 | NP_001007146.1 | 9 | 1272 | ||
XM_008772037.1 | XP_008770259.1 | 3 | 180 | |||
GenBank mRNA | BC085789.1 | - | 9 | 1272 | 7108824 |
