siRNA Details
Gene Symbol: Sun1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007147.1 | NP_001007148.1 | 10 | 1242 | ||
XM_006248941.2 | XP_006249003.1 | 14 | 1776 | |||
XM_006248942.2 | XP_006249004.1 | 14 | 1776 | |||
XM_006248943.1 | XP_006249005.1 | 15 | 1856 | |||
XM_006248945.1 | XP_006249007.1 | 14 | 1746 | |||
XM_006248946.1 | XP_006249008.1 | 15 | 1828 | |||
XM_006248947.2 | XP_006249009.1 | 14 | 2982 | |||
XM_006248948.2 | XP_006249010.1 | 13 | 1665 | |||
XM_006248949.2 | XP_006249011.1 | 13 | 1578 | |||
XM_006248951.2 | XP_006249013.1 | 12 | 1467 | |||
XM_006248952.2 | XP_006249014.1 | 11 | 1401 | |||
XM_006248953.2 | XP_006249015.1 | 10 | 1302 | |||
XM_008768982.1 | XP_008767204.1 | 16 | 2006 | |||
XM_008768983.1 | XP_008767205.1 | 14 | 1904 | |||
XM_008768984.1 | XP_008767206.1 | 13 | 1624 | |||
GenBank mRNA | BC085844.1 | - | 10 | 1242 | 7135918 |
