siRNA Details
Gene Symbol: FAM131B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031690.2 | NP_001026860.2 | 5 | 440 | ||
NM_001278297.1 | NP_001265226.1 | 4 | 346 | |||
NM_014690.4 | NP_055505.3 | 5 | 333 | |||
XM_005250073.3 | XP_005250130.1 | 5 | 654 | |||
XM_006716186.2 | XP_006716249.1 | 5 | 432 | |||
GenBank mRNA | AB018316.1 | - | 4 | 228 | ||
AK124557.1 | - | 5 | 388 | |||
AK291470.1 | - | 5 | 305 | |||
BC045611.1 | - | 5 | 445 | 5300056 | ||
BC050543.1 | - | 5 | 308 | 5768309 | ||
DA226407.1 | - | 5 | 432 | |||
DA334424.1 | - | 4 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
