siRNA Details
Gene Symbol: Fblim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007554.1 | NP_001007555.1 | 9 | 1333 | ||
XM_008764273.1 | XP_008762495.1 | 9 | 3557 | |||
XM_008764274.1 | XP_008762496.1 | 8 | 2720 | |||
XM_008764275.1 | XP_008762497.1 | 9 | 2789 | |||
XM_008764276.1 | XP_008762498.1 | 9 | 2940 | |||
GenBank mRNA | BC086398.1 | - | 9 | 1333 | 7303205 |
