siRNA Details
Gene Symbol: Dhrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007621.1 | NP_001007622.1 | 5 | 810 | ||
XM_006251946.2 | XP_006252008.1 | 5 | 675 | |||
XM_006251947.2 | XP_006252009.1 | 6 | 743 | |||
XM_006251948.2 | XP_006252010.1 | 6 | 721 | |||
GenBank mRNA | BC079263.1 | - | 5 | 810 | 7133513 |
