siRNA Details
Gene Symbol: Ascc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007632.2 | NP_001007633.1 | 9 | 956 | ||
XM_006256404.2 | XP_006256466.1 | 9 | 1150 | |||
XM_006256405.2 | XP_006256467.1 | 9 | 1147 | |||
XM_006256407.2 | XP_006256469.2 | 9 | 1036 | |||
XM_006256408.2 | XP_006256470.1 | 9 | 1105 | |||
XM_006256409.1 | XP_006256471.1 | 8 | 891 | |||
XM_006256410.2 | XP_006256472.1 | 9 | 1102 | |||
XM_008772881.1 | XP_008771103.1 | 8 | 888 | |||
GenBank mRNA | BP504504.1 | - | 5 | 447 | ||
FQ226270.1 | - | 6 | 603 |
