siRNA Details
Gene Symbol: Mocs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007633.2 | NP_001007634.1 | 6 | 1296 | ||
NM_001162413.1 | NP_001155885.1 | 7 | 1418 | |||
XM_006231985.2 | XP_006232047.1 | 7 | 1364 | |||
XM_006231986.2 | XP_006232048.1 | 6 | 1261 | |||
GenBank mRNA | BC079181.1 | - | 7 | 1384 | 7130858 |
