siRNA Details
Gene Symbol: Mocs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007633.2 | NP_001007634.1 | 2, 3 | 276 | ||
NM_001162413.1 | NP_001155885.1 | 3, 4 | 398 | |||
GenBank mRNA | BC079181.1 | - | 3, 4 | 364 | 7130858 | |
CO554692.1 | - | 3, 4 | 398 | |||
FM123325.1 | - | 2, 3 | 230 | |||
FQ230755.1 | - | 2, 3 | 199 | |||
FQ235152.1 | - | 3, 4 | 326 |
