siRNA Details
Gene Symbol: Rhno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW143602.1 | - | 3 | 384 | ||
BC079219.1 | - | 3 | 770 |
Gene Symbol: LOC100362315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761349.1 | XP_008759571.1 | 1 | 393 |

More Information
Other Species Targeted

