siRNA Details
Gene Symbol: PTGER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198714.1 | NP_942007.1 | 5 | 1474 | ||
NM_198716.1 | NP_942009.1 | 4 | 1382 | |||
NM_198717.1 | NP_942010.1 | 3 | 1355 | |||
NR_028292.1 | 6 | 1551 | ||||
NR_028293.1 | 5 | 1524 | ||||
NR_028294.1 | 6 | 1620 | ||||
XM_011541810.2 | XP_011540112.1 | 4 | 1453 | |||
GenBank mRNA | AK290634.1 | - | 5 | 1340 | ||
BC024229.1 | - | 4 | 1392 | 4660371 | ||
D38298.1 | - | 5 | 1474 | |||
D38300.1 | - | 4 | 1382 | |||
D38301.1 | - | 3 | 1355 | |||
D86097.1 | - | 6 | 1551 | |||
D86098.1 | - | 5 | 1524 | |||
L27489.1 | - | 3 | 1341 | |||
S68875.1 | - | 1 | 47 | |||
U13214.1 | - | 4 | 1207 | |||
U13215.1 | - | 3 | 1180 | |||
X83857.1 | - | 5 | 1450 | |||
X83859.1 | - | 3 | 1331 | |||
X83861.1 | - | 4 | 1358 | |||
X83863.1 | - | 6 | 1596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
