siRNA Details
Gene Symbol: Mmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007673.1 | NP_001007674.1 | 4 | 476 | ||
XM_006247126.1 | XP_006247188.1 | 4 | 514 | |||
XM_006247127.2 | XP_006247189.1 | 4 | 1336 | |||
GenBank mRNA | AF540876.1 | - | 4 | 375 | ||
BC081965.1 | - | 4 | 476 | 7125235 |
