siRNA Details
Gene Symbol: Abhd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007680.1 | NP_001007681.1 | 9 | 1033 | ||
XM_006251766.2 | XP_006251828.1 | 9 | 2312 | |||
XM_006251767.2 | XP_006251829.1 | 9 | 1844 | |||
XM_006251768.2 | XP_006251830.1 | 9 | 1009 | |||
GenBank mRNA | BC083826.1 | - | 9 | 1033 | 7112208 | |
FQ219067.1 | - | 4 | 311 |
