siRNA Details
Gene Symbol: Tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007701.1 | NP_001007702.1 | 1726 | |||
GenBank mRNA | BC083851.1 | - | 1726 | 7123274 | ||
FQ209507.1 | - | 1 | 71 | |||
FQ219758.1 | - | 1 | 67 | |||
FQ223598.1 | - | 1600 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
