siRNA Details
Gene Symbol: Nudt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007733.1 | NP_001007734.1 | 2 | 165 | ||
XM_008771875.1 | XP_008770097.1 | 2 | 184 | |||
GenBank mRNA | BC079176.1 | - | 2 | 165 | 7130276 |
