siRNA Details
Gene Symbol: RGD1359158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007737.1 | NP_001007738.1 | 4 | 1007 | ||
GenBank mRNA | BC079346.1 | - | 4 | 1007 | 7107242 | |
BC093390.1 | - | 4 | 998 | 7388009 | ||
FQ215421.1 | - | 3 | 811 |
