siRNA Details
Gene Symbol: Cyb561d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007753.1 | NP_001007754.1 | 3 | 709 | ||
XM_006243804.2 | XP_006243866.1 | 4 | 362 | |||
GenBank mRNA | BC082072.1 | - | 3 | 709 | 7133078 | |
FQ213574.1 | - | 3 | 400 |
