siRNA Details
Gene Symbol: Rassf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007754.1 | NP_001007755.1 | 1 | 50 | ||
GenBank mRNA | BC079237.1 | - | 1 | 50 | 7132629 | |
FQ233012.1 | - | 1 | 90 |

More Information
Other Species Targeted

Gene Ontology Categories:
