siRNA Details
Gene Symbol: Cir1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007799.1 | NP_001007800.1 | 8 | 515 | ||
GenBank mRNA | BC085869.1 | - | 8 | 585 | 7191148 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025854, BC094283, AK005900, AK006260, AK131674, and others.
Gene Ontology Categories:
