siRNA Details
Gene Symbol: Clpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007803.1 | NP_001007804.1 | 8, 9 | 1056 | ||
XM_006243250.1 | XP_006243312.1 | 7, 8 | 1259 | |||
GenBank mRNA | BC085867.1 | - | 8, 9 | 1182 | 7190926 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011802, NM_001044389, BC061153, AJ276991, AF134983, and others.
Gene Ontology Categories:
