siRNA Details
Gene Symbol: Mtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008286.1 | NP_001008287.1 | 9, 10 | 790 | ||
XM_006234370.2 | XP_006234432.1 | 9, 10 | 621 | |||
GenBank mRNA | BC086360.1 | - | 9, 10 | 790 | 7303884 | |
FQ215122.1 | - | 9, 10 | 758 | |||
FQ215507.1 | - | 9, 10 | 785 | |||
FQ215582.1 | - | 9, 10 | 808 |
